Gene Transcripts

Gene Symbol
ABTB1
Entrez Gene ID
80325
Gene Name
ankyrin repeat and BTB domain containing 1
Gene Aliases
BPOZ, BTB3, BTBD21, EF1ABP, PP2259
Chromosome Location
Chr. 3: 127672928 - 127680926 on Build GRCh38
UniGene ID
Hs.107812
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032548.3 NP_115937.1 5 513
NM_172027.2 NP_742024.1 5 502
NR_033429.1 5 467
XM_006713769.3 XP_006713832.1 5 536
XM_011513213.1 XP_011511515.1 5 436
XM_017007285.1 XP_016862774.1 5 482
XM_017007286.1 XP_016862775.1 5 464
XM_017007287.1 XP_016862776.1 5 473
XM_017007288.1 XP_016862777.1 5 523
GenBank mRNA AB053324.1 - 5 492
AB053325.1 - 5 481
AB053326.1 - 5 446
AF297986.1 - 5 459
AF447886.1 - 4 603
AK090422.1 - 1 880
AK315148.1 - 5 482
BC011858.2 - 5 500 4309664
BF664138.1 - 5 499

View Full Product Details