Gene Transcripts

Gene Symbol
KIRREL2
Entrez Gene ID
84063
Gene Name
kin of IRRE like 2 (Drosophila)
Gene Aliases
FILTRIN, NEPH3, NLG1
Chromosome Location
Chr. 19: 35851635 - 35867146 on Build GRCh38
UniGene ID
Hs.145729
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032123.6 NP_115499.5 7 902
NM_199179.3 NP_954648.3 6 752
NM_199180.3 NP_954649.3 7 1044
XM_011527362.1 XP_011525664.1 8 1104
XM_011527363.1 XP_011525665.1 8 1085
XM_011527365.1 XP_011525667.1 7 954
XM_011527366.1 XP_011525668.1 6 1215
XM_017027348.1 XP_016882837.1 7 1611
GenBank mRNA AB593116.1 - 7 1008
AK307865.1 - 7 1365
AL136654.1 - 7 1028
AY305301.1 - 7 846
AY305302.1 - 6 696
AY358742.1 - 7 1030
BC064925.1 - 10 967 6141879

View Full Product Details