Gene Transcripts

Gene Symbol
UQCRC1
Entrez Gene ID
7384
Gene Name
ubiquinol-cytochrome c reductase core protein I
Gene Aliases
D3S3191, QCR1, UQCR1
Chromosome Location
Chr. 3: 48598999 - 48609665 on Build GRCh38
UniGene ID
Hs.119251
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003365.2 NP_003356.2 5 572
GenBank mRNA AK129903.1 - 5 552
AK300694.1 - 5 507
AK302840.1 - 5 597
AK313090.1 - 5 542
BC009586.1 - 5 543 4096628
D26485.1 - 5 534
L16842.1 - 5 944

View Full Product Details