Gene Transcripts

Gene Symbol
FBXL12
Entrez Gene ID
54850
Gene Name
F-box and leucine rich repeat protein 12
Gene Aliases
Fbl12
Chromosome Location
Chr. 19: 9810267 - 9820527 on Build GRCh38
UniGene ID
Hs.12439
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316936.1 NP_001303865.1 2 1613
NM_001316937.1 NP_001303866.1 3 1686
NM_001316938.1 NP_001303867.1 4 1756
NM_001316939.1 NP_001303868.1 4 1730
NM_001316940.1 NP_001303869.1 3 1608
NM_001316941.1 NP_001303870.1 2 1639
NM_001316942.1 NP_001303871.1 4 1784
NM_017703.2 NP_060173.1 3 1712
XM_006722782.3 XP_006722845.2 2 1463
XM_017026912.1 XP_016882401.1 3 1588
GenBank mRNA AK000195.1 - 3 1664
AK093760.1 - 3 1659
BC001586.2 - 3 1616 2966439
BX647073.1 - 2 1549

View Full Product Details