Gene Transcripts

Gene Symbol
PIP4K2B
Entrez Gene ID
8396
Gene Name
phosphatidylinositol-5-phosphate 4-kinase type 2 beta
Gene Aliases
PI5P4KB, PIP5K2B, PIP5KIIB, PIP5KIIbeta, PIP5P4KB
Chromosome Location
Chr. 17: 38765691 - 38799965 on Build GRCh38
UniGene ID
Hs.260603
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003559.4 NP_003550.1 2, 3 747
XM_011525326.2 XP_011523628.1 2, 3 807
XM_011525327.2 XP_011523629.1 3, 4 326
XM_011525329.2 XP_011523631.1 2, 3 807
XM_017025196.1 XP_016880685.1 3, 4 324
XM_017025197.1 XP_016880686.1 1, 2 57
XM_017025199.1 XP_016880688.1 2, 3 157
GenBank mRNA BC027459.1 - 2, 3 364
BT019614.1 - 2, 3 266
U85245.1 - 2, 3 746

View Full Product Details