Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001006622.2 | NP_001006623.1 | 6 | 804 | |
NM_001006623.2 | NP_001006624.1 | 6 | 804 | ||
NM_018383.4 | NP_060853.3 | 6 | 804 | ||
XM_005263697.2 | XP_005263754.1 | 6 | 804 | ||
XM_011511436.1 | XP_011509738.1 | 6 | 833 | ||
XM_017004435.1 | XP_016859924.1 | 6 | 804 | ||
XM_017004436.1 | XP_016859925.1 | 6 | 804 | ||
GenBank mRNA | AA142986.1 | - | 1 | 15 | |
AB044749.1 | - | 6 | 764 | ||
AK002156.1 | - | 6 | 779 | ||
AK223319.1 | - | 6 | 781 | ||
BC005401.1 | - | 6 | 788 | ||
BC013990.2 | - | 6 | 779 | ||
BC068484.1 | - | 6 | 779 |