Gene Transcripts

Gene Symbol
PARL
Entrez Gene ID
55486
Gene Name
presenilin associated rhomboid like
Gene Aliases
PRO2207, PSARL, PSARL1, PSENIP2, RHBDS1
Chromosome Location
Chr. 3: 183826488 - 183884908 on Build GRCh38
UniGene ID
Hs.478469
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037639.2 NP_001032728.1 5 637
NM_001324436.1 NP_001311365.1 5 637
NM_001324437.1 NP_001311366.1 5 637
NM_001324438.1 NP_001311367.1 5 637
NM_018622.6 NP_061092.3 5 637
NR_136893.1 5 693
XM_005247582.4 XP_005247639.1 5 632
XM_017006800.1 XP_016862289.1 5 632
XM_017006801.1 XP_016862290.1 5 630
XM_017006802.1 XP_016862291.1 5 630
XM_017006803.1 XP_016862292.1 5 684
GenBank mRNA AF197937.1 - 5 575
AK024765.1 - 5 609
BC003653.1 - 5 624 3537438
BC014058.2 - 5 595 3937238

View Full Product Details