Gene Transcripts

Gene Symbol
CYSLTR2
Entrez Gene ID
57105
Gene Name
cysteinyl leukotriene receptor 2
Gene Aliases
CYSLT2, CYSLT2R, GPCR21, HG57, HPN321, KPG_011, PSEC0146, hGPCR21
Chromosome Location
Chr. 13: 48653711 - 48709362 on Build GRCh38
UniGene ID
Hs.253706
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308465.1 NP_001295394.1 6 1010
NM_001308467.1 NP_001295396.1 6 987
NM_001308468.1 NP_001295397.1 4 808
NM_001308469.1 NP_001295398.1 4 785
NM_001308470.1 NP_001295399.1 3 718
NM_001308471.1 NP_001295400.1 2 617
NM_001308476.1 NP_001295405.1 5 881
NM_020377.3 NP_065110.1 5 858
GenBank mRNA AB038269.1 - 1 572
AB041947.1 - 1 643
AK291739.1 - 5 858
AY853711.1 - 4 537
BC069160.1 - 1 312 7216884
BC096832.1 - 1 425 6971946

View Full Product Details