Gene Transcripts

Gene Symbol
SCAF4
Entrez Gene ID
57466
Gene Name
SR-related CTD associated factor 4
Gene Aliases
SFRS15, SRA4
Chromosome Location
Chr. 21: 31671000 - 31732118 on Build GRCh38
UniGene ID
Hs.17255
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145444.1 NP_001138916.1 10 1644
NM_001145445.1 NP_001138917.1 11 1689
NM_020706.2 NP_065757.1 11 1689
XM_005261017.2 XP_005261074.1 10 1493
XM_006724035.2 XP_006724098.1 11 1670
XM_006724036.2 XP_006724099.1 11 1670
XM_017028415.1 XP_016883904.1 11 1647
XM_017028416.1 XP_016883905.1 11 1532
XM_017028417.1 XP_016883906.1 10 1493
GenBank mRNA AB032998.1 - 5 677
AF023142.1 - 11 1293
AK025748.1 - 6 708
AK308406.1 - 10 1495
AL117417.1 - 6 697
AL834304.1 - 11 1469
BC014921.1 - 11 1666 3907610
BC043353.1 - 11 1646 6019414
BC052286.1 - 11 1520 5442227
BC064990.1 - 11 1520 5432277

View Full Product Details