Gene Transcripts

Gene Symbol
VPS11
Entrez Gene ID
55823
Gene Name
VPS11, CORVET/HOPS core subunit
Gene Aliases
END1, HLD12, PEP5, RNF108, hVPS11
Chromosome Location
Chr. 11: 119067752 - 119081978 on Build GRCh38
UniGene ID
Hs.234282
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290185.1 NP_001277114.1 5 1114
NM_021729.5 NP_068375.3 5 899
XM_011542899.2 XP_011541201.1 5 857
GenBank mRNA AB027508.2 - 5 869
AF258558.1 - 3 400
AF308800.1 - 5 833
AK023279.1 - 5 870
AK056701.1 - 4 726
AK223158.1 - 5 859
AK302973.1 - 5 1046
AL834218.1 - 5 846
BC065563.1 - 5 833 6144541

View Full Product Details