Gene Transcripts

Gene Symbol
GPR88
Entrez Gene ID
54112
Gene Name
G protein-coupled receptor 88
Gene Aliases
COCPMR, STRG
Chromosome Location
Chr. 1: 100538172 - 100542027 on Build GRCh38
UniGene ID
Hs.170053
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022049.2 NP_071332.2 2 2315
GenBank mRNA AB042410.1 - 2 2348

View Full Product Details