Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018518.4 | NP_060988.3 | 3 | 283 | |
NM_182751.2 | NP_877428.1 | 3 | 283 | ||
XM_011519538.2 | XP_011517840.1 | 3 | 253 | ||
GenBank mRNA | AB042719.1 | - | 3 | 250 | |
AK055695.1 | - | 3 | 256 | ||
AK292701.1 | - | 3 | 258 | ||
AL136840.1 | - | 2 | 249 | ||
BC101727.1 | - | 3 | 252 | 8069233 | |
BC143490.1 | - | 3 | 248 | 9051998 | |
DA024424.1 | - | 3 | 283 |