Gene Transcripts

Gene Symbol
TEX14
Entrez Gene ID
56155
Gene Name
testis expressed 14, intercellular bridge forming factor
Gene Aliases
CT113
Chromosome Location
Chr. 17: 58556677 - 58692055 on Build GRCh38
UniGene ID
Hs.390221
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001201457.1 NP_001188386.1 4 498
NM_031272.4 NP_112562.3 4 498
NM_198393.3 NP_938207.2 4 498
XM_011525029.2 XP_011523331.1 4 477
XM_017024860.1 XP_016880349.1 3 262
XM_017024861.1 XP_016880350.1 4 502
GenBank mRNA AK125896.1 - 4 488
AK308299.1 - 4 458
AL834143.1 - 4 497
BC040526.1 - 4 466 4831308
BG718877.1 - 4 470
DB086121.1 - 4 522

View Full Product Details