Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001205263.1 | NP_001192192.1 | 3 | 702 | |
NM_012415.3 | NP_036547.1 | 5 | 679 | ||
GenBank mRNA | AF112481.1 | - | 5 | 628 | |
AK290437.1 | - | 5 | 649 | ||
AK307516.1 | - | 3 | 375 | ||
BC001965.2 | - | 5 | 621 | 3452509 | |
BC020668.1 | - | 6 | 692 |