Gene Transcripts
- Gene Symbol
- SNX5
- Entrez Gene ID
- 27131
- Gene Name
- sorting nexin 5
- Gene Aliases
- Chromosome Location
- Chr. 20: 17941596 - 17968991 on Build GRCh38
- UniGene ID
- Hs.316890
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282454.1 |
NP_001269383.1
|
4, 5 |
637 |
|
NM_014426.3 |
NP_055241.1
|
4, 5 |
765 |
|
NM_152227.2 |
NP_689413.1
|
5, 6 |
720 |
|
GenBank mRNA |
AF121855.1 |
- |
5, 6 |
576 |
|
AK001793.1 |
- |
4, 5 |
482 |
|
AK054634.1 |
- |
4, 5 |
1201 |
|
AK129505.1 |
- |
4, 5 |
482 |
|
AK223308.1 |
- |
4, 5 |
450 |
|
AK296947.1 |
- |
5, 6 |
589 |
|
AK308017.1 |
- |
5, 6 |
721 |
|
BC062638.1 |
- |
4, 5 |
422 |
|
BC093623.1 |
- |
4, 5 |
691 |
7939468 |
BC093980.1 |
- |
4, 5 |
691 |
7939825 |
BC143274.1 |
- |
4, 5 |
691 |
9051776 |
View Full Product Details