Gene Transcripts

Gene Symbol
CNTN6
Entrez Gene ID
27255
Gene Name
contactin 6
Gene Aliases
NB3
Chromosome Location
Chr. 3: 978021 - 1403610 on Build GRCh38
UniGene ID
Hs.387300
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289080.1 NP_001276009.1 5 942
NM_001289081.1 NP_001276010.1 4 805
NM_014461.3 NP_055276.1 5 951
XM_005265058.3 XP_005265115.1 6 1074
XM_011533590.2 XP_011531892.1 5 598
XM_011533591.2 XP_011531893.1 4 913
XM_017006171.1 XP_016861660.1 10 1501
XM_017006172.1 XP_016861661.1 5 1076
XM_017006173.1 XP_016861662.1 5 690
XM_017006174.1 XP_016861663.1 4 565
XM_017006175.1 XP_016861664.1 3 342
XM_017006176.1 XP_016861665.1 5 690
XM_017006177.1 XP_016861666.1 5 690
GenBank mRNA AB003592.1 - 5 664
AK294786.1 - 4 805
AK307814.1 - 6 691
AK314220.1 - 5 665
BC113118.1 - 5 577 40036214

View Full Product Details