Gene Transcripts

Gene Symbol
SNX9
Entrez Gene ID
51429
Gene Name
sorting nexin 9
Gene Aliases
SDP1, SH3PX1, SH3PXD3A, WISP
Chromosome Location
Chr. 6: 157716046 - 157945077 on Build GRCh38
UniGene ID
Hs.191213
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016224.4 NP_057308.1 18 3430
XM_005267015.2 XP_005267072.1 20 3508
GenBank mRNA AB209736.1 - 11 7398
AF190158.1 - 1 1283
AJ420452.1 - 1 243
AJ420512.1 - 1 1234
AK127345.1 - 15 3285

View Full Product Details