Gene Transcripts

Gene Symbol
ANAPC7
Entrez Gene ID
51434
Gene Name
anaphase promoting complex subunit 7
Gene Aliases
APC7
Chromosome Location
Chr. 12: 110372900 - 110403730 on Build GRCh38
UniGene ID
Hs.719935
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001137664.1 NP_001131136.1 10 1599
NM_016238.2 NP_057322.2 10 1599
XM_005253903.3 XP_005253960.1 9 1465
XM_011538454.2 XP_011536756.1 10 1827
XM_017019425.1 XP_016874914.1 9 1374
XM_017019426.1 XP_016874915.1 10 1594
XM_017019427.1 XP_016874916.1 10 2393
XM_017019428.1 XP_016874917.1 9 1482
GenBank mRNA AF191340.1 - 10 1496
AK021809.1 - 10 1578
AL137586.1 - 4 653
AY007104.1 - 3 969
BC002941.2 - 5 743
BC009498.2 - 8 1169
BC017316.2 - 6 945
BC040130.1 - 10 1514
BC098264.1 - 10 1495
BC098295.1 - 10 1495
BC098362.1 - 10 1495
BC099732.1 - 10 1495
BC111798.1 - 10 1495
BC141848.1 - 10 1549 8860346
BC148236.2 - 10 1496 40024981

View Full Product Details