Gene Transcripts

Gene Symbol
CLIC5
Entrez Gene ID
53405
Gene Name
chloride intracellular channel 5
Gene Aliases
DFNB102, DFNB103, MST130, MSTP130
Chromosome Location
Chr. 6: 45898451 - 46129809 on Build GRCh38
UniGene ID
Hs.485489
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114086.1 NP_001107558.1 4 931
NM_001256023.1 NP_001242952.1 4 695
NM_016929.4 NP_058625.2 4 695
XM_011514692.2 XP_011512994.1 4 931
XM_011514694.2 XP_011512996.1 4 931
XM_017010953.1 XP_016866442.1 5 1176
GenBank mRNA AF176920.1 - 4 494
AF216941.1 - 4 645
AK075144.1 - 4 634
AK075163.1 - 4 546
AK097048.1 - 4 931
AK222544.1 - 4 663
AK223130.1 - 4 666
AK314416.1 - 4 511
AY032602.1 - 4 825
BC035968.1 - 4 663 4611102
DQ679794.1 - 4 663

View Full Product Details