Gene Transcripts

Gene Symbol
LANCL1
Entrez Gene ID
10314
Gene Name
LanC like 1
Gene Aliases
GPR69A, p40
Chromosome Location
Chr. 2: 210431249 - 210477618 on Build GRCh38
UniGene ID
Hs.13351
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136574.1 NP_001130046.1 3 193
NM_001136575.1 NP_001130047.1 3 188
NM_006055.2 NP_006046.1 3 282
XM_005246243.2 XP_005246300.1 3 303
GenBank mRNA AK294674.1 - 3 220
AK309295.1 - 3 173
AK312381.1 - 3 171
BC028685.2 - 3 193 4829574
CR541777.1 - 2 118
DC321883.1 - 3 188
DC324843.1 - 3 283
Y11395.1 - 3 222

View Full Product Details