Gene Transcripts

Gene Symbol
TOMM34
Entrez Gene ID
10953
Gene Name
translocase of outer mitochondrial membrane 34
Gene Aliases
HTOM34P, TOM34, URCC3
Chromosome Location
Chr. 20: 44942130 - 44960520 on Build GRCh38
UniGene ID
Hs.517066
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006809.4 NP_006800.2 5 804
XM_005260254.2 XP_005260311.1 5 737
XM_011528501.1 XP_011526803.1 5 623
GenBank mRNA AB085681.1 - 5 729
AK000594.1 - 5 723
AK222952.1 - 5 711
AK302128.1 - 5 633
AK307655.1 - 5 745
BC001763.1 - 5 717 3544124
BC007423.2 - 5 717 2960090
BC014907.1 - 5 694 3922502
BT020008.1 - 5 664
BT020009.1 - 5 664
CR456765.1 - 5 664
U58970.1 - 5 695

View Full Product Details