Gene Transcripts

Gene Symbol
PLA2G16
Entrez Gene ID
11145
Gene Name
phospholipase A2 group XVI
Gene Aliases
AdPLA, H-REV107-1, HRASLS3, HREV107, HREV107-1, HREV107-3, HRSL3
Chromosome Location
Chr. 11: 63574462 - 63616926 on Build GRCh38
UniGene ID
Hs.502775
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128203.1 NP_001121675.1 5 768
NM_007069.3 NP_009000.2 4 1052
XM_006718426.1 XP_006718489.1 5 970
XM_011544741.1 XP_011543043.1 4 954
GenBank mRNA X92814.1 - 4 1035

View Full Product Details