Gene Transcripts

Gene Symbol
PAXIP1
Entrez Gene ID
22976
Gene Name
PAX interacting protein 1
Gene Aliases
CAGF28, CAGF29, PACIP1, PAXIP1L, PTIP, TNRC2
Chromosome Location
Chr. 7: 154943690 - 155003393 on Build GRCh38
UniGene ID
Hs.443881
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005249539.1 XP_005249596.1 5 365
XM_011515983.1 XP_011514285.1 4 265
XM_017011881.1 XP_016867370.1 5 1538
GenBank mRNA AK123600.1 - 5 365

View Full Product Details