Gene Transcripts

Gene Symbol
FRAT2
Entrez Gene ID
23401
Gene Name
frequently rearranged in advanced T-cell lymphomas 2
Gene Aliases
Chromosome Location
Chr. 10: 97332497 - 97334701 on Build GRCh38
UniGene ID
Hs.140720
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012083.2 NP_036215.1 1 1128
GenBank mRNA AB045118.1 - 1 1128
BC020165.1 - 1 1103 3629702

View Full Product Details