Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286715.1 | NP_001273644.1 | 6 | 774 | |
NM_001286717.1 | NP_001273646.1 | 6 | 774 | ||
NM_015239.2 | NP_056054.2 | 7 | 839 | ||
XM_005251847.3 | XP_005251904.1 | 7 | 785 | ||
XM_005251848.2 | XP_005251905.1 | 7 | 963 | ||
XM_005251849.3 | XP_005251906.1 | 6 | 709 | ||
XM_011518418.2 | XP_011516720.1 | 7 | 785 | ||
XM_011518421.2 | XP_011516723.1 | 7 | 785 | ||
XM_017014544.1 | XP_016870033.1 | 7 | 1330 | ||
XM_017014545.1 | XP_016870034.1 | 7 | 785 | ||
XM_017014546.1 | XP_016870035.1 | 6 | 672 | ||
XM_017014547.1 | XP_016870036.1 | 6 | 709 | ||
XM_017014548.1 | XP_016870037.1 | 7 | 690 | ||
XM_017014549.1 | XP_016870038.1 | 6 | 709 | ||
XM_017014550.1 | XP_016870039.1 | 7 | 785 | ||
GenBank mRNA | AB028958.2 | - | 7 | 697 | |
AK001544.1 | - | 8 | 639 | ||
AK295774.1 | - | 6 | 774 | ||
AK299506.1 | - | 3 | 334 | ||
BC060815.1 | - | 7 | 804 | 30344122 |