Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001035260.2 | NP_001030337.1 | 3 | 318 | |
NM_001318944.1 | NP_001305873.1 | 4 | 424 | ||
NM_001318945.1 | NP_001305874.1 | 4 | 394 | ||
NM_004896.4 | NP_004887.2 | 3 | 318 | ||
GenBank mRNA | AF054179.1 | - | 3 | 272 | |
AF175266.1 | - | 3 | 272 | ||
AK022992.1 | - | 3 | 248 | ||
AK297142.1 | - | 4 | 363 | ||
AK309021.1 | - | 4 | 372 | ||
AK315496.1 | - | 3 | 292 | ||
BC022505.1 | - | 3 | 305 | 4799288 | |
BQ048905.1 | - | 2 | 187 | ||
BQ068008.1 | - | 2 | 199 | ||
CD358963.1 | - | 444 | |||
DA498726.1 | - | 3 | 318 |