Gene Transcripts

Gene Symbol
OXSR1
Entrez Gene ID
9943
Gene Name
oxidative stress responsive 1
Gene Aliases
OSR1
Chromosome Location
Chr. 3: 38165051 - 38255488 on Build GRCh38
UniGene ID
Hs.475970
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005109.2 NP_005100.1 18 2474
XM_005265638.2 XP_005265695.1 19 2615
XM_011534331.2 XP_011532633.1 18 2526
XM_017007601.1 XP_016863090.1 17 2281
XM_017007602.1 XP_016863091.1 20 2801
GenBank mRNA AB017642.1 - 18 2474
AB029024.1 - 18 2227
BC008726.2 - 18 2457 3163379

View Full Product Details