Gene Transcripts
- Gene Symbol
- TNFSF15
- Entrez Gene ID
- 9966
- Gene Name
- tumor necrosis factor superfamily member 15
- Gene Aliases
- TL1, TL1A, TNLG1B, VEGI, VEGI192A
- Chromosome Location
- Chr. 9: 114784635 - 114806128 on Build GRCh38
- UniGene ID
- Hs.23349
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001204344.1 |
NP_001191273.1
|
1, 2 |
118 |
|
NM_005118.3 |
NP_005109.2
|
3, 4 |
411 |
|
GenBank mRNA |
AF520785.1 |
- |
3, 4 |
417 |
|
AK291642.1 |
- |
3, 4 |
322 |
|
AK313721.1 |
- |
3, 4 |
386 |
|
AY434464.1 |
- |
1, 2 |
118 |
|
BC069435.1 |
- |
3, 4 |
366 |
7262219 |
BC074940.2 |
- |
3, 4 |
335 |
30915494 |
BC074941.2 |
- |
3, 4 |
335 |
30915271 |
BC104462.1 |
- |
3, 4 |
410 |
40030800 |
BC104463.1 |
- |
3, 4 |
411 |
40030803 |
View Full Product Details