Gene Transcripts

Gene Symbol
SMC4
Entrez Gene ID
10051
Gene Name
structural maintenance of chromosomes 4
Gene Aliases
CAP-C, CAPC, SMC-4, SMC4L1
Chromosome Location
Chr. 3: 160399304 - 160434962 on Build GRCh38
UniGene ID
Hs.58992
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002800.2 NP_001002800.1 3 694
NM_001288753.1 NP_001275682.1 3 619
NM_005496.3 NP_005487.3 2 511
XM_006713459.3 XP_006713522.1 3 464
XM_011512311.2 XP_011510613.1 3 582
XM_011512312.2 XP_011510614.1 2 203
XM_017005491.1 XP_016860980.1 1 29
GenBank mRNA AB019987.1 - 3 355
AK002200.1 - 3 348
AK128143.1 - 3 273
AK225437.1 - 3 348
AK315070.1 - 3 348
AL136877.1 - 2 475
AL833949.1 - 2 1402
BC001557.1 - 3 330
BC065259.1 - 3 526
BC070161.1 - 3 348
BC106033.1 - 3 366

View Full Product Details