Gene Transcripts

Gene Symbol
DDX39A
Entrez Gene ID
10212
Gene Name
DEAD-box helicase 39A
Gene Aliases
BAT1, BAT1L, DDX39, DDXL, URH49
Chromosome Location
Chr. 19: 14408798 - 14419383 on Build GRCh38
UniGene ID
Hs.311609
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005804.3 NP_005795.2 4, 5 555
NR_046366.2 4, 5 555
XM_006722606.3 XP_006722669.1 4, 5 495
XM_011527620.1 XP_011525922.1 4, 5 574
XM_011527621.2 XP_011525923.1 3, 4 441
GenBank mRNA AB254849.1 - 3, 4 437
AK301847.1 - 5, 6 638
BC001009.2 - 4, 5 492 2820926
BC032128.2 - 4, 5 501 5017293
U90426.1 - 4, 5 532

View Full Product Details