Gene Transcripts

Gene Symbol
PNMA1
Entrez Gene ID
9240
Gene Name
paraneoplastic Ma antigen 1
Gene Aliases
MA1
Chromosome Location
Chr. 14: 73711783 - 73714425 on Build GRCh38
UniGene ID
Hs.194709
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006029.4 NP_006020.4 1 1340
GenBank mRNA AF320308.1 - 1 825
AK290980.1 - 1 677
AL834327.1 - 1 73
BC039577.1 - 1 643 4431875
CR457112.1 - 1 554

View Full Product Details