Gene Transcripts
- Gene Symbol
- HIST1H2AG
- Entrez Gene ID
- 8969
- Gene Name
- histone cluster 1, H2ag
- Gene Aliases
- H2A.1b, H2A/p, H2AFP, H2AG, pH2A/f
- Chromosome Location
- Chr. 6: 27133038 - 27133535 on Build GRCh38
- UniGene ID
- Hs.51011
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC016677.1 |
- |
1 |
2117 |
|
L19778.1 |
- |
1 |
2128 |
|
View Full Product Details