Gene Transcripts

Gene Symbol
MPHOSPH9
Entrez Gene ID
10198
Gene Name
M-phase phosphoprotein 9
Gene Aliases
MPP-9, MPP9
Chromosome Location
Chr. 12: 123152324 - 123244014 on Build GRCh38
UniGene ID
Hs.577404
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022782.3 NP_073619.3 16 2741
NR_103517.1 16 2480
XM_011537740.2 XP_011536042.1 16 2645
XM_011537741.2 XP_011536043.1 16 2645
XM_017018673.1 XP_016874162.1 16 2739
XM_017018675.1 XP_016874164.1 15 2547
XM_017018676.1 XP_016874165.1 15 2489
XM_017018677.1 XP_016874166.1 16 2645
XM_017018678.1 XP_016874167.1 15 2465
XM_017018679.1 XP_016874168.1 17 2678
XM_017018680.1 XP_016874169.1 15 2587
XM_017018681.1 XP_016874170.1 15 2500
GenBank mRNA AK023016.1 - 11 1464
AK091181.1 - 12 2070
AK096541.1 - 16 2353
AL096751.1 - 10 1401
BC130440.1 - 12 2023 40146471
BC144016.1 - 11 1897
BC144017.1 - 11 1675

View Full Product Details