Gene Transcripts
- Gene Symbol
- UMOD
- Entrez Gene ID
- 7369
- Gene Name
- uromodulin
- Gene Aliases
- ADMCKD2, FJHN, HNFJ, HNFJ1, MCKD2, THGP, THP
- Chromosome Location
- Chr. 16: 20333051 - 20356301 on Build GRCh38
- UniGene ID
- Hs.654425
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001008389.2 |
NP_001008390.1
|
5, 6 |
1418 |
|
NM_001278614.1 |
NP_001265543.1
|
6, 7 |
1580 |
|
NM_003361.3 |
NP_003352.2
|
5, 6 |
1481 |
|
XM_011545934.1 |
XP_011544236.1
|
5, 6 |
1418 |
|
XM_011545935.1 |
XP_011544237.1
|
6, 7 |
1477 |
|
XM_011545936.1 |
XP_011544238.1
|
6, 7 |
1414 |
|
XM_011545937.1 |
XP_011544239.1
|
6, 7 |
1304 |
|
XM_011545938.1 |
XP_011544240.1
|
5, 6 |
1336 |
|
XM_011545940.1 |
XP_011544242.1
|
5, 6 |
1354 |
|
XM_017023641.1 |
XP_016879130.1
|
5, 6 |
1418 |
|
GenBank mRNA |
AK055722.1 |
- |
6, 7 |
1184 |
|
AK091961.1 |
- |
6, 7 |
1412 |
|
AK096043.1 |
- |
6, 7 |
1061 |
|
AK127643.1 |
- |
6, 7 |
914 |
|
AK127648.1 |
- |
6, 7 |
1177 |
|
BC035975.1 |
- |
6, 7 |
1167 |
4556427 |
M15881.1 |
- |
5, 6 |
1357 |
|
M17778.1 |
- |
4, 5 |
1294 |
|
View Full Product Details