Gene Transcripts

Gene Symbol
DYSF
Entrez Gene ID
8291
Gene Name
dysferlin
Gene Aliases
FER1L1, LGMD2B, MMD1
Chromosome Location
Chr. 2: 71453155 - 71686763 on Build GRCh38
UniGene ID
Hs.252180
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130455.1 NP_001123927.1 11 1155
NM_001130976.1 NP_001124448.1 11 1387
NM_001130977.1 NP_001124449.1 11 1387
NM_001130978.1 NP_001124450.1 11 1387
NM_001130979.1 NP_001124451.1 12 1480
NM_001130980.1 NP_001124452.1 12 1480
NM_001130981.1 NP_001124453.1 12 1480
NM_001130982.1 NP_001124454.1 12 1248
NM_001130983.1 NP_001124455.1 11 1155
NM_001130984.1 NP_001124456.1 11 1155
NM_001130985.1 NP_001124457.1 12 1248
NM_001130986.1 NP_001124458.1 11 1155
NM_001130987.1 NP_001124459.1 12 1248
NM_003494.3 NP_003485.1 11 1387
XM_005264584.4 XP_005264641.1 12 1263
XM_005264585.4 XP_005264642.1 12 1948
GenBank mRNA AF075575.1 - 11 1384
BC171803.1 - 11 1360
DQ267935.1 - 11 1155
EU515155.1 - 12 1381
EU515156.1 - 11 1288
EU515157.1 - 11 1288
EU515158.1 - 12 1381
EU515159.1 - 11 1288
EU515160.1 - 12 1381
EU515161.1 - 12 1248
EU515162.1 - 11 1155
EU515163.1 - 11 1155
EU515164.1 - 12 1248
EU515165.1 - 11 1155
EU515166.1 - 12 1248

View Full Product Details