Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003608.3 | NP_003599.2 | 2 | 1325 | |
GenBank mRNA | AK313797.1 | - | 2 | 1300 | |
BC035633.1 | - | 2 | 1297 | 5218256 | |
BC071715.1 | - | 2 | 1259 | 5180030 | |
EU883574.1 | - | 1 | 767 |