Gene Transcripts

Gene Symbol
THBS3
Entrez Gene ID
7059
Gene Name
thrombospondin 3
Gene Aliases
TSP3
Chromosome Location
Chr. 1: 155195588 - 155209180 on Build GRCh38
UniGene ID
Hs.169875
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252607.1 NP_001239536.1 10 1392
NM_001252608.1 NP_001239537.1 9 1059
NM_007112.4 NP_009043.1 11 1419
NR_045553.1 10 1403
XM_006711498.3 XP_006711561.1 11 2978
XM_011509926.2 XP_011508228.1 11 2978
XM_011509927.2 XP_011508229.1 10 2951
XM_011509928.2 XP_011508230.1 11 2885
XM_011509929.1 XP_011508231.1 11 1374
XM_011509930.2 XP_011508232.1 11 2978
XM_011509931.1 XP_011508233.1 11 1324
XM_011509932.2 XP_011508234.1 11 1257
XM_011509933.1 XP_011508235.1 12 1606
XM_011509934.1 XP_011508236.1 11 1361
XM_011509935.1 XP_011508237.1 11 1517
XM_011509936.1 XP_011508238.1 10 1258
XM_017002206.1 XP_016857695.1 10 1331
GenBank mRNA AK074796.1 - 8 755
AK223278.1 - 11 1333
AK298592.1 - 9 995
AK298803.1 - 8 887
AK298914.1 - 2 180
AK314337.1 - 10 1343
BC018786.1 - 2 222
BC113846.1 - 10 1321 40008346
BC113847.1 - 11 1337 40008343
L38969.1 - 11 1335

View Full Product Details