Gene Transcripts

Gene Symbol
B9D1
Entrez Gene ID
27077
Gene Name
B9 domain containing 1
Gene Aliases
B9, EPPB9, MKS9, MKSR1
Chromosome Location
Chr. 17: 19335372 - 19378193 on Build GRCh38
UniGene ID
Hs.309467
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243473.2 NP_001230402.1 4 375
NM_001243475.2 NP_001230404.1 4 571
NM_001321214.1 NP_001308143.1 4 468
NM_001321215.1 NP_001308144.1 4 468
NM_001321216.1 NP_001308145.1 4 468
NM_001321217.1 NP_001308146.1 4 468
NM_001321218.1 NP_001308147.1 4 468
NM_001321219.1 NP_001308148.1 4 468
NM_015681.4 NP_056496.1 4 468
XM_005256607.2 XP_005256664.1 4 440
XM_005256610.1 XP_005256667.1 4 443
XM_017024452.1 XP_016879941.1 7 2832
GenBank mRNA AB030506.1 - 4 457
AK293815.1 - 4 441
AK297782.1 - 4 358
AK309320.1 - 4 560
BC002944.2 - 4 399 3953287
BI833202.1 - 4 463

View Full Product Details