Gene Transcripts

Gene Symbol
FNDC8
Entrez Gene ID
54752
Gene Name
fibronectin type III domain containing 8
Gene Aliases
Chromosome Location
Chr. 17: 35121612 - 35130732 on Build GRCh38
UniGene ID
Hs.120369
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017559.2 NP_060029.1 1 216
GenBank mRNA AK225743.1 - 1 181
AK313775.1 - 1 249
AL133054.1 - 1 167
BC024002.2 - 1 197 4821771
BI829497.1 - 1 217

View Full Product Details