Gene Transcripts

Gene Symbol
APPL2
Entrez Gene ID
55198
Gene Name
adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2
Gene Aliases
DIP13B
Chromosome Location
Chr. 12: 105173297 - 105236230 on Build GRCh38
UniGene ID
Hs.506603
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001251904.1 NP_001238833.1 17 1790
NM_001251905.1 NP_001238834.1 17 1815
NM_018171.3 NP_060641.2 17 1772
XM_006719472.1 XP_006719535.1 17 1709
XM_011538530.2 XP_011536832.1 17 3242
XM_011538531.2 XP_011536833.1 17 2905
XM_011538532.2 XP_011536834.1 17 4231
XM_017019551.1 XP_016875040.1 17 2177
XM_017019552.1 XP_016875041.1 17 2887
XM_017019553.1 XP_016875042.1 18 4485
XM_017019554.1 XP_016875043.1 17 1691
GenBank mRNA AK001521.2 - 17 1592
AK290562.1 - 17 1625
AK293783.1 - 17 1564
AK296610.1 - 17 1571
AK297100.1 - 17 1643
AK310377.1 - 17 1560
AY113704.1 - 17 1772
BC033731.1 - 17 1676 5168590

View Full Product Details