Gene Transcripts

Gene Symbol
CTC1
Entrez Gene ID
80169
Gene Name
CTS telomere maintenance complex component 1
Gene Aliases
AAF-132, AAF132, C17orf68, CRMCC, tmp494178
Chromosome Location
Chr. 17: 8224821 - 8248095 on Build GRCh38
UniGene ID
Hs.156055
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025099.5 NP_079375.3 17 2857
NR_046431.1 17 2752
XM_006721577.3 XP_006721640.1 16 2712
XM_006721578.3 XP_006721641.1 17 2841
XM_011524010.2 XP_011522312.1 17 2736
XM_011524011.2 XP_011522313.1 16 2606
GenBank mRNA AK025823.1 - 7 1000
AK091077.1 - 17 2808
AK225375.1 - 7 1000
AK310636.1 - 6 1026
AL831955.1 - 17 2699
BC026057.2 - 8 998
BC110373.1 - 9 1224
BC111783.1 - 17 2808 40054070
BF798182.1 - 1 39
BX406692.2 - 4 416
DQ451688.1 - 17 2798
FJ790670.1 - 17 2798

View Full Product Details