Gene Transcripts

Gene Symbol
CTC1
Entrez Gene ID
80169
Gene Name
CTS telomere maintenance complex component 1
Gene Aliases
AAF-132, AAF132, C17orf68, CRMCC, tmp494178
Chromosome Location
Chr. 17: 8224821 - 8248095 on Build GRCh38
UniGene ID
Hs.156055
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025099.5 NP_079375.3 22 3555
NR_046431.1 21 3385
XM_006721577.3 XP_006721640.1 21 3410
XM_006721578.3 XP_006721641.1 20 3308
XM_011524010.2 XP_011522312.1 22 3434
XM_011524011.2 XP_011522313.1 21 3304
GenBank mRNA AK025823.1 - 12 1698
AK091077.1 - 22 3506
AK225375.1 - 12 1698
AL831955.1 - 21 3332
BC026057.2 - 13 1696
BC110373.1 - 14 1922
BC111783.1 - 22 3506 40054070
DQ451688.1 - 22 3496
FJ790670.1 - 22 3496

View Full Product Details