Gene Transcripts

Gene Symbol
ZNRF1
Entrez Gene ID
84937
Gene Name
zinc and ring finger 1, E3 ubiquitin protein ligase
Gene Aliases
NIN283
Chromosome Location
Chr. 16: 74999017 - 75110994 on Build GRCh38
UniGene ID
Hs.427284
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032268.4 NP_115644.1 5 2029
GenBank mRNA AF378524.1 - 5 1701
AL390144.1 - 1 3008
AL834440.1 - 6 1506

View Full Product Details