Gene Transcripts

Gene Symbol
TCHP
Entrez Gene ID
84260
Gene Name
trichoplein keratin filament binding
Gene Aliases
TpMs
Chromosome Location
Chr. 12: 109880667 - 109918069 on Build GRCh38
UniGene ID
Hs.410924
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143852.1 NP_001137324.1 2 325
NM_032300.4 NP_115676.1 2 386
XM_011538836.2 XP_011537138.1 2 661
XM_011538837.2 XP_011537139.1 2 234
XM_017020024.1 XP_016875513.1 2 325
XM_017020025.1 XP_016875514.1 2 349
GenBank mRNA AK092736.1 - 2 325
AK301262.1 - 2 303
AY007230.1 - 2 387
BC004285.1 - 2 306 3617849
BC022838.1 - 2 615

View Full Product Details