Gene Transcripts

Gene Symbol
COQ10A
Entrez Gene ID
93058
Gene Name
coenzyme Q10A
Gene Aliases
Chromosome Location
Chr. 12: 56266858 - 56270966 on Build GRCh38
UniGene ID
Hs.4096
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099337.1 NP_001092807.1 5 1151
NM_144576.3 NP_653177.3 5 1395
GenBank mRNA AK057003.1 - 5 1311
AK057014.1 - 4 1112
AY358861.1 - 5 1083
BC002435.2 - 4 972
BC026922.1 - 5 1151
BC047444.1 - 5 1123
BC073923.1 - 5 1279 5188138

View Full Product Details