Gene Transcripts

Gene Symbol
CRHR1-IT1
Entrez Gene ID
147081
Gene Name
CRHR1 intronic transcript 1
Gene Aliases
C17orf69
Chromosome Location
Chr. 17: 45638975 - 45646229 on Build GRCh38
UniGene ID
Hs.128813
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026905.2 2 2220
GenBank mRNA AK057897.1 - 2 980
AK090656.1 - 1 1699
BC069230.1 - 2 1941

View Full Product Details