Gene Transcripts

Gene Symbol
OTOGL
Entrez Gene ID
283310
Gene Name
otogelin like
Gene Aliases
C12orf64, DFNB84B
Chromosome Location
Chr. 12: 80130377 - 80379090 on Build GRCh38
UniGene ID
Hs.723594
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173591.3 NP_775862.3 58 7849
XM_005268802.3 XP_005268859.1 62 8650
XM_011538191.1 XP_011536493.1 59 7926
XM_011538192.2 XP_011536494.1 56 7867
XM_011538193.1 XP_011536495.1 56 7786
GenBank mRNA AK075060.1 - 6 1452
AK096852.1 - 12 2127

View Full Product Details