Gene Transcripts

Gene Symbol
ANKRD33
Entrez Gene ID
341405
Gene Name
ankyrin repeat domain 33
Gene Aliases
C12orf7, PANKY
Chromosome Location
Chr. 12: 51888009 - 51891721 on Build GRCh38
UniGene ID
Hs.433492
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130015.1 NP_001123487.1 6 1463
NM_001304459.1 NP_001291388.1 6 1726
NM_001304460.1 NP_001291389.1 5 1337
NM_182608.3 NP_872414.3 5 1637
GenBank mRNA AF517120.1 - 1 220
AK125517.1 - 5 1686
BX537908.1 - 6 1238
BX640656.1 - 6 1238
BX647684.1 - 2 2937
BX649168.1 - 6 1472

View Full Product Details