Gene Transcripts

Gene Symbol
PRKCG
Entrez Gene ID
5582
Gene Name
protein kinase C gamma
Gene Aliases
PKC-gamma, PKCC, PKCG, SCA14
Chromosome Location
Chr. 19: 53882184 - 53907647 on Build GRCh38
UniGene ID
Hs.631564
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316329.1 NP_001303258.1 3 545
NM_002739.4 NP_002730.1 3 545
GenBank mRNA AB209671.1 - 3 264
AF345987.1 - 3 429
AK296412.1 - 3 553
AK296434.1 - 3 532
AK302958.1 - 3 530
AK303741.1 - 2 64
AK312302.1 - 3 507
BC047876.1 - 3 420 5764695
BI757265.1 - 3 518
DC318717.1 - 3 545
M13977.1 - 3 429

View Full Product Details