Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001206838.1 | NP_001193767.1 | 9 | 1406 | |
NM_001206839.1 | NP_001193768.1 | 9 | 1406 | ||
NM_002851.2 | NP_002842.2 | 9 | 1406 | ||
XM_005250519.1 | XP_005250576.1 | 9 | 1406 | ||
XM_017012477.1 | XP_016867966.1 | 10 | 1453 | ||
GenBank mRNA | M93426.1 | - | 9 | 1158 | |
U88967.1 | - | 9 | 1346 |